Learn More
c-jun, p Thr93, p Thr91 Antibody (C-J 4C4/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-2748-0.025mg
 View more versions of this product
Product Code. 18530421
  • € 190.00 / 25µg
Description
c-jun Monoclonal specifically detects c-jun in Human, Rat samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
c-jun (p Thr91, p Thr93)
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000
JUN
C-Jun phosphorylated at T91 and T93
0.025 mg
Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
3725
Human
Purified
Western Blot
C-J 4C4/1
Unconjugated
Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian)
Mouse
Protein A purified
RUO
Primary
This recognises c-Jun phosphorylated at T91 and T93, or just at T91 but not at T93 alone. Cross-reacts with Human and Rat. Not yet tested in other species.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.