missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-jun, p Thr93, p Thr91 Antibody (C-J 4C4/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00
Specifications
| Antigen | c-jun (p Thr91, p Thr93) |
|---|---|
| Clone | C-J 4C4/1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
| Applications | Western Blot |
Description
c-jun Monoclonal specifically detects c-jun in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
| c-jun (p Thr91, p Thr93) | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
| Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
| JUN | |
| IgG1 | |
| Protein A purified | |
| This recognises c-Jun phosphorylated at T91 and T93, or just at T91 but not at T93 alone. Cross-reacts with Human and Rat. Not yet tested in other species. |
| C-J 4C4/1 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 3725 | |
| C-Jun phosphorylated at T91 and T93 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title