Learn More
C17orf57 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27603-1-AP-150UL
 View more versions of this product
Product Code. 18094815
  • € 410.00 / 150µL
Description
The C17orf57 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C17orf57. This antibody recognizes mouse antigen. The C17orf57 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
C17orf57
450 μg/mL
Rabbit
Primary
Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
C17orf57, EFCAB13
150 μL
124989
Store at -20°C. Stable for one year after shipment.