missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf57 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | C17orf57 |
|---|---|
| Concentration | 450 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18094815
|
Proteintech
27603-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18035205
|
Proteintech
27603-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The C17orf57 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C17orf57. This antibody recognizes mouse antigen. The C17orf57 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| C17orf57 | |
| Immunohistochemistry, ELISA | |
| Rabbit | |
| 124989 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 450 μg/mL | |
| Liquid | |
| Mouse | |
| C17orf57, EFCAB13 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title