missing translation for 'onlineSavingsMsg'
Learn More

C19orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen C19orf43
Concentration 0.23 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898974
View Documents
Proteintech
19420-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888974
View Documents
Proteintech
19420-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The antibody was raised from the C19orf43 protein. A band at ∽28kd was detected in Western Blot with human placenta, 293 cell, and A431 cell lysates.

C19orf43, also named as MGC2803 and fSAP18, is a protein coding gene. Exoribonuclease that is part of the telomerase RNA 3′ end processing complex and which has the ability to all four unpaired RNA nucleotides from 5′ end or 3′ end with higher efficiency for purine bases.
Specifications

Specifications

C19orf43
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Rat, Human, Mouse
Q9BQ61
288920, 68544, 79002
C19orf43 Fusion Protein Ag13747
Primary
-20°C
C19orf43
0.23 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
C19orf43, fSAP18
C19orf43, Trir
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.