Learn More
C1QBP Rabbit anti-Human, Unconjugated, Clone: 3K20, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80490-1-RR-100UL
Description
The C1QBP antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human C1QBP. This antibody recognizes human, mouse, rat antigen. The C1QBP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| C1QBP | |
| 3K20 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| 100 μL | |
| 708 | |
| IgG |