missing translation for 'onlineSavingsMsg'
Learn More

C1QBP Rabbit anti-Human, Unconjugated, Clone: 3K20, Proteintech

Rabbit Recombinant Antibody

€ 149.00 - € 439.00

Specifications

Antigen C1QBP
Clone 3K20
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17551506
View Documents
Proteintech
80490-1-RR-20UL
20 μL
€ 149.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17541506
View Documents
Proteintech
80490-1-RR-100UL
100 μL
€ 439.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The C1QBP antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human C1QBP. This antibody recognizes human, mouse, rat antigen. The C1QBP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.

Specifications

Specifications

C1QBP
Western Blot, Immunohistochemistry, Immunofluorescence
Rabbit
708
Primary
3K20
Unconjugated
Human, Mouse, Rat
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.