Learn More
uvex C300 Wet Cut Protection Gloves
Shop All Uvex Products
Click to view available options
Size:
8
10
Unit Size:
Each
Description
- Soft-grip coating ensures mechanical abrasion resistance in wet applications
- Bamboo TwinFlex™ Technology provides a high level of cut resistance
- Highly flexible with good tactile sensitivity
- Silicone-free according to the imprint test
Specifications
Specifications
| Coating Material | HPE |
| Color | Green, Black |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | HPPE, Polyamide, Bamboo Viscose |
| Product Type | Cut-resistant Gloves |
| Size | 10 |
| EN388 Standard | 4X42C |
| Series | C300 Wet |