missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf58 Antibody (1G1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50604-0.025ml
This item is not returnable.
View return policy
Description
C6orf58 Monoclonal specifically detects C6orf58 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| C6orf58 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| chromosome 6 open reading frame 58, hypothetical protein LOC352999, MGC72003 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 352999 | |
| Zebrafish | |
| Purified |
| Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1G1 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C6ORF58 | |
| GST-C6orf58 fusion protein | |
| 0.025 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction