missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf58 Antibody (1G1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00 - € 409.00
Specifications
| Antigen | C6orf58 |
|---|---|
| Clone | 1G1 |
| Concentration | 1.0 mg/mL |
| Dilution | Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18157448
|
Novus Biologicals
NBP2-50604 |
0.1 mL |
€ 409.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695448
|
Novus Biologicals
NBP2-50604-0.025ml |
0.025 mL |
€ 190.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C6orf58 Monoclonal specifically detects C6orf58 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| C6orf58 | |
| 1.0 mg/mL | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Zebrafish | |
| chromosome 6 open reading frame 58, hypothetical protein LOC352999, MGC72003 | |
| C6ORF58 | |
| IgG1 | |
| Protein A purified |
| 1G1 | |
| Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 352999 | |
| GST-C6orf58 fusion protein | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title