missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99567-50ul
This item is not returnable.
View return policy
Description
CACNG1 Polyclonal antibody specifically detects CACNG1 in Human samples. It is validated for Western Blot
Specifications
| CACNG1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CACNLGDihydropyridine-sensitive L-type, skeletal muscle calcium channel subunit gamma, calcium channel, voltage-dependent, gamma subunit 1, L-type calcium channel gamma polypeptide, neuronal dihydropyridine-sensitive calcium channel gamma subunit, voltage-dependent calcium channel gamma-1 subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CACNG1. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 786 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction