missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 227.00 - € 307.65
Specifications
| Antigen | CACNG1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668005
|
Novus Biologicals
NBP2-99567-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621806
|
Novus Biologicals
NBP2-99567-50ul |
50 μg |
€ 227.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CACNG1 Polyclonal antibody specifically detects CACNG1 in Human samples. It is validated for Western BlotSpecifications
| CACNG1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CACNLGDihydropyridine-sensitive L-type, skeletal muscle calcium channel subunit gamma, calcium channel, voltage-dependent, gamma subunit 1, L-type calcium channel gamma polypeptide, neuronal dihydropyridine-sensitive calcium channel gamma subunit, voltage-dependent calcium channel gamma-1 subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CACNG1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 786 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title