Learn More
Novus Biologicals™ CALHM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30945-25ug
 View more versions of this product
Product Code. 30067158
  • € 213.00 / 25µg
Description
CALHM1 Polyclonal antibody specifically detects CALHM1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CALHM1
Polyclonal
Western Blot : 1-5 μg/mL, ELISA
calcium homeostasis modulator 1, FAM26Ccalcium homeostasis modulator protein 1, family with sequence similarity 26, member C, MGC39514, MGC39617, Protein FAM26C
Synthetic peptide derived from human CALHM1 protein.
25 μg
Alzheimers Research, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
255022
Store at -20°C. Avoid freeze/thaw cycles.
IgG
ELISA, Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation