missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody (AF18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB300-518-0.025ml
This item is not returnable.
View return policy
Description
Calnexin Monoclonal specifically detects Calnexin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Calnexin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:1000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20 | |
| CANX | |
| Focus human hepatoma cell lysate. | |
| 0.025 mL | |
| Vision | |
| 821 | |
| Human | |
| Purified |
| Western Blot | |
| AF18 | |
| Unconjugated | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Calnexin (AF18) | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction