missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody (AF18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
€ 226.00
Specifications
| Antigen | Calnexin |
|---|---|
| Clone | AF18 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:1000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20 |
| Applications | Western Blot |
Description
Calnexin Monoclonal specifically detects Calnexin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Calnexin | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Vision | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| CANX | |
| IgG1 | |
| Protein A purified | |
| Calnexin (AF18) |
| AF18 | |
| Western Blot 1:100-1:1000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 821 | |
| Focus human hepatoma cell lysate. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title