Learn More
Calponin-1 Mouse, Clone: rCNN1/832, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1264-MSM6-P1
This item is not returnable.
View return policy
Specifications
| Calponin-1 | |
| Cytoplasmic | |
| rCNN1/832 | |
| Unconjugated | |
| XP_005259798 | |
| Mouse | |
| 34kDa | |
| Protein A/G Purified | |
| RUO | |
| 1264 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| CNN1 | |
| Recombinant full-length human CNN1 protein | |
| Myoepithelial cells in breast ducts | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |