Learn More
Calponin-1 Mouse, Clone: rCNN1/832, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | Calponin-1 |
|---|---|
| Clone | rCNN1/832 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15959837
|
EnquireBio
1264-MSM6-P0 |
20 μg | |||||||
|
15969837
|
EnquireBio
1264-MSM6-P1 |
100 μg |
€ 554.40 € 478.00 / 100µg Save € 76.40 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Calponin-1 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 1264 | |
| Recombinant full-length human CNN1 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 34kDa |
| rCNN1/832 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| XP_005259798 | |
| CNN1 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic | |
| Myoepithelial cells in breast ducts |