missing translation for 'onlineSavingsMsg'
Learn More

Calretinin Antibody, Novus Biologicals™

Chicken Polyclonal Antibody

Brand:  Novus Biologicals NBP2-50029

 View more versions of this product

Product Code. 18686378

  • € 503.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calretinin Polyclonal antibody specifically detects Calretinin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calretinin
Polyclonal
Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry Free-Floating
P22676
Chicken
IgY purified
RUO
Primary
Human, Mouse, Rat, Pig, Bovine
Purified
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Unconjugated
Supplied as an aliquot of IgY preparation
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR
Recombinant full length human Calretinin. [UniProt# P22676]
0.1 mL
Cellular Markers
794
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.