missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
€ 183.00 - € 503.00
Specifications
| Antigen | Calretinin |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry Free-Floating |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686378
|
Novus Biologicals
NBP2-50029 |
0.1 mL |
€ 503.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666798
|
Novus Biologicals
NBP2-50029SS |
0.025 mL |
€ 183.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calretinin Polyclonal antibody specifically detects Calretinin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-FloatingSpecifications
| Calretinin | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| Unconjugated | |
| Chicken | |
| Cellular Markers | |
| Supplied as an aliquot of IgY preparation | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| Recombinant full length human Calretinin. [UniProt# P22676] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry Free-Floating | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine | |
| P22676 | |
| 794 | |
| IgY | |
| IgY purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title