Learn More
Honeywell KCL™ Camatril™ 730 Gloves
Product Code. 10328154
Click to view available options
Size:
7
8
9
10
Unit Size:
10 pairs
Each
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10328154
Brand: Honeywell KCL™ 073007141E
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Honeywell KCL™ Camatril™ 730 Gloves are a top-quality protective glove made of high-quality nitrile, making them useful for a wide range of applications.
  • Top-quality nitrile
  • Large spectrum of chemical resistance
  • Versatile glove suitable for numerous fields
  • Good flexibility
  • Anatomical design and excellent fit
  • Good sensitivity
  • Flock lining for added comfort
  • Approved for food handling and processing according to EU 1935/2004
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EC Category PPE 3, suitable for food handling according to EU 1935/2004
Coating Material Nitrile
Color Green
Cut Resistance Rating 0
Material Nitrile
Size 7
Thickness (Metric) 0.4 mm
Lining Flock Lined
Puncture Rating 1
Resistant To Resistance to permeation by chemicals: AJL
Tear Rating 0
Length (Metric) 300-320 mm