Learn More
Honeywell KCL™ Camatril™ 730 Gloves
Click to view available options
Size:
7
8
9
10
Unit Size:
10 pairs
Each
Description
- Top-quality nitrile
- Large spectrum of chemical resistance
- Versatile glove suitable for numerous fields
- Good flexibility
- Anatomical design and excellent fit
- Good sensitivity
- Flock lining for added comfort
- Approved for food handling and processing according to EU 1935/2004
Specifications
Specifications
| Certifications/Compliance (Europe) | EC Category PPE 3, suitable for food handling according to EU 1935/2004 |
| Coating Material | Nitrile |
| Color | Green |
| Cut Resistance Rating | 0 |
| Material | Nitrile |
| Size | 7 |
| Thickness (Metric) | 0.4 mm |
| Lining | Flock Lined |
| Puncture Rating | 1 |
| Resistant To | Resistance to permeation by chemicals: AJL |
| Show More |