Learn More
Thermo Scientific™ Thermo Scientific™ 10 mL and 12 mL Screw Top Transport Tubes and Caps
GREENER_CHOICE
Product Code. 17185311
Click to view available options
Volume (Metric):
10 mL
12 mL
90 mL
118 mL
Closure Type:
Flip Top
Not Included
Screw Cap
Color:
Amber
Clear
Natural
Closure Color:
Blue
Clear
Green
Not Included
Red
White
Yellow
Unit Size:
Pack of 1000
Pack of 2000
Pack of 250
Pack of 400
Product Code. 17185311
Brand: Thermo Scientific™ S40600
Only null left
Add to basket
Add to basket
Choose 10 mL and 12 mL screw top tubes for sample collection and transport. The tubes are manufactured from a food grade polypropylene resin for visual clarity and sample security.
Thermo Scientific™ 0 mL and 12 mL Screw Top Transport Tubes are made from virgin, clarified polypropylene resin for visual clarity and sample security.
  • Freestanding
  • Non-sterile tubes
10 mL
  • Graduations in 1 mL increments up to 8 mL
  • 0.59 O.D. x 3.38 in. L (15 x 86 mm)
  • Round bottom with skirt
12 mL
  • Etched fill lines at 5 mL and 8 mL
  • 0.59 O.D. x 3.77 in. L (15 x 96 mm)
  • Conical bottom with skirt
  • Available in amber color for light sensitive samples
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Yellow
Color Amber
Diameter (Metric) Outer 15 mm
Material Polypropylene
Sterility Non-sterile
Volume (Metric) 12 mL
No. per Case 1000/Cs.
Style Skirted, conical bottom
Closure Material Polyethylene
Closure Type Screw Cap
Height (Metric) 96 mm
Type Sample Vial