missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1026, G10260, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, AISI 1026, G10260, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Titanium/Calcium/Aluminium/Niobium/Oxygen/Cobalt/Arsenic/Nitrogen/Phosphorus/Tin/Molybdenum/Vanadium/Nickel/Chromium/Copper/Silicon/Carbon/Manganese/Magnesium/Lead/Tungsten/Zirconium/Antimony |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Titanium 0.0009 %/Calcium 0.0013 %/Aluminium 0.002 %/Niobium 0.002 %/Oxygen 0.0044 %/Cobalt 0.0069 %/Arsenic 0.0073 %/Nitrogen 0.0094 %/Phosphorus 0.0094 %/Tin 0.01 %/Molybdenum 0.023 %/Vanadium 0.027 %/Nickel 0.068 %/Chromium 0.121 %/Copper 0.186 %/Silicon 0.233 %/Carbon 0.267 %/Manganese 0.686 %/Magnesium (0.0005) %/Lead (0.001) %/Tungsten (0.001) %/Zirconium (0.001) %/Antimony (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction