missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1030, G10300, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, AISI 1030, G10300, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Lead/Niobium/Titanium/Tungsten/Calcium/Aluminium/Antimony/Oxygen/Arsenic/Cobalt/Phosphorus/Nitrogen/Tin/Molybdenum/Vanadium/Nickel/Chromium/Copper/Silicon/Carbon/Manganese |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0002 %/Lead 0.001 %/Niobium 0.001 %/Titanium 0.001 %/Tungsten 0.001 %/Calcium 0.002 %/Aluminium 0.003 %/Antimony 0.003 %/Oxygen 0.005 %/Arsenic 0.006 %/Cobalt 0.006 %/Phosphorus 0.009 %/Nitrogen 0.0107 %/Tin 0.011 %/Molybdenum 0.036 %/Vanadium 0.042 %/Nickel 0.079 %/Chromium 0.14 %/Copper 0.243 %/Silicon 0.27 %/Carbon 0.32 %/Manganese 0.67 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction