missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Resulfurized, AISI 1117, G11170, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, Resulfurized, AISI 1117, G11170, 100g
Specifications
Specifications
| Chemical Name or Material | Boron/Lead/Calcium/Titanium/Niobium/Antimony/Aluminium/Oxygen/Cobalt/Arsenic/Nitrogen/Tin/Phosphorus/Vanadium/Molybdenum/Nickel/Chromium/Carbon/Silicon/Copper/Manganese/Zirconium/Tungsten/Bismuth/Magnesium/Tantalum/Selenium/Zinc |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0007 %/Lead 0.001 %/Calcium 0.0012 %/Titanium 0.0014 %/Niobium 0.0024 %/Antimony 0.003 %/Aluminium 0.0032 %/Oxygen 0.005 %/Cobalt 0.008 %/Arsenic 0.0085 %/Nitrogen 0.0093 %/Tin 0.0109 %/Phosphorus 0.0157 %/Vanadium 0.0255 %/Molybdenum 0.0269 %/Nickel 0.082 %/Chromium 0.105 %/Carbon 0.193 %/Silicon 0.239 %/Copper 0.253 %/Manganese 1.19 %/Zirconium (<0.004) %/Tungsten (<0.007) %/Bismuth (<0.013) %/Magnesium (0.0003) %/Tantalum (0.0004) %/Selenium (0.0006) %/Zinc (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction