Learn More
Cardiac Troponin T Mouse, anti-Mouse, CoraLite Plus 488, Clone: 4E2D8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68300-20UL
 View more versions of this product
Product Code. 18029485
  • € 145.00 / 20µL
Description
The Cardiac Troponin T antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cardiac Troponin T. This antibody recognizes mouse, rat, rabbit, pig, chicken antigen. The Cardiac Troponin T antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Cardiac Troponin T
4E2D8
CoraLite Plus 488
Mouse
Primary
Mouse, Rat, Rabbit, Pig, Chicken
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
Cardiac muscle troponin T, CMH2, CMPD2, cTnT, RCM3, TNNT2, TnTC, troponin T type 2 (cardiac), Troponin T, cardiac muscle
20 μL
7139
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.