Learn More
Cardiac Troponin T Mouse, anti-Mouse, CoraLite Plus 488, Clone: 4E2D8, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | Cardiac Troponin T |
|---|---|
| Clone | 4E2D8 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18029485
|
Proteintech
CL488-68300-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18085215
|
Proteintech
CL488-68300-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Cardiac Troponin T antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cardiac Troponin T. This antibody recognizes mouse, rat, rabbit, pig, chicken antigen. The Cardiac Troponin T antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Cardiac Troponin T | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 7139 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 4E2D8 | |
| Immunofluorescence | |
| Liquid | |
| Mouse, Rat, Rabbit, Pig, Chicken | |
| Cardiac muscle troponin T, CMH2, CMPD2, cTnT, RCM3, TNNT2, TnTC, troponin T type 2 (cardiac), Troponin T, cardiac muscle | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.