Learn More
Cardiac Troponin T Mouse, anti-Mouse, CoraLite Plus 488, Clone: 4E2D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68300-100UL
Description
The Cardiac Troponin T antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cardiac Troponin T. This antibody recognizes mouse, rat, rabbit, pig, chicken antigen. The Cardiac Troponin T antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| Cardiac Troponin T | |
| 4E2D8 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Mouse, Rat, Rabbit, Pig, Chicken | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| Cardiac muscle troponin T, CMH2, CMPD2, cTnT, RCM3, TNNT2, TnTC, troponin T type 2 (cardiac), Troponin T, cardiac muscle | |
| 100 μL | |
| 7139 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |