missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASRegistryNumber1t Iron, Low Alloy, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
CASRegistryNumber1t Iron, Low Alloy, 50g
Specifications
Specifications
| Chemical Name or Material | Bismuth/Tellurium/Zirconium/Boron/Selenium/Tungsten/Lead/Nitrogen/Zinc/Niobium/Arsenic/Antimony/Tin/Cobalt/Aluminium/Vanadium/Phosphorus/Titanium/Copper/Molybdenum/Manganese/Chromium/Nickel/Silicon/Carbon |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Bismuth (0.002) %/Tellurium (0.002) %/Zirconium (0.002) %/Boron (0.0055) %/Selenium (0.007) %/Tungsten (0.007) %/Lead (0.01) %/Nitrogen (0.01) %/Zinc (0.01) %/Niobium (0.018) %/Arsenic (0.021) %/Antimony (0.03) %/Tin (0.035) %/Cobalt (0.048) %/Aluminium (0.06) %/Vanadium (0.074) %/Phosphorus (0.09) %/Titanium (0.094) %/Copper (0.43) %/Molybdenum (0.53) %/Manganese (0.76) %/Chromium (0.95) %/Nickel (1.15) %/Silicon (1.92) %/Carbon (2.77) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction