Learn More
Fisherbrand™ Catalyst Tygon E-Food Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
€ 224.00 - € 354.00
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | ||||
15521345
|
Fisherbrand™
|
1.6 mm | 1.6 mm |
€ 282.00
Pack of 12 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
||||
15511345
|
Fisherbrand™
|
0.6 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15531345
|
Fisherbrand™
|
3.2 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15541345
|
Fisherbrand™
|
4.8 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15551345
|
Fisherbrand™
|
6.4 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15561345
|
Fisherbrand™
|
8 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15571345
|
Fisherbrand™
|
4.8 mm | 2.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15591345
|
Fisherbrand™
|
8 mm | 2.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15380058
|
Fisherbrand™
|
9.5 mm | 2.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Available in Platinum-Cured Silicone, TPEs, and PVC
- Ensures optimal amount of tubing is in pump head
Specifications
Stopped tubing assembly |