Learn More
MP Biomedicals™ Catch Tubes
Product Code. 15656104 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15656104 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15656104 Supplier MP Biomedicals™ Supplier No. 116560211
Only null left
Add to basket
Add to basket
Catch Tubes are used during the silica-based purification procedure to eliminate accidental transfer of silica particles.
  • Included in FastDNA SPIN Kits and in the GENECLEAN SPIN Kit
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Quantity 50 Pk.
Product Type Catch Tubes
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.