missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin alpha 1 Antibody (8B6C1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61722-0.025ml
This item is not returnable.
View return policy
Description
Catenin alpha 1 Monoclonal specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Catenin alpha 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13 | |
| Mouse | |
| 100 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 8B6C1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CTNNA1 | |
| Purified recombinant fragment of human CTNNA1 (AA: 371-574) expressed in E. Coli. | |
| Protein G purified | |
| Cancer, Cellular Markers, Core ESC Like Genes, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers | |
| 1495 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction