missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin alpha 1 Antibody (8B6C1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | Catenin alpha 1 |
|---|---|
| Clone | 8B6C1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217974
|
Novus Biologicals
NBP2-61722 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18617896
|
Novus Biologicals
NBP2-61722-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Catenin alpha 1 Monoclonal specifically detects Catenin alpha 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Catenin alpha 1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Alpha E-catenin, alpha-catenin, alpha-E-catenin, alphaE-catenin, Cadherin-associated protein, CAP102, catenin (cadherin-associated protein), alpha 1 (102kD), catenin (cadherin-associated protein), alpha 1, 102kDa, catenin alpha-1, FLJ36832, FLJ52416, Renal carcinoma antigen NY-REN-13 | |
| CTNNA1 | |
| IgG1 | |
| Protein G purified |
| 8B6C1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| Cancer, Cellular Markers, Core ESC Like Genes, Cytoskeleton Markers, Phospho Specific, Stem Cell Markers | |
| 1495 | |
| Purified recombinant fragment of human CTNNA1 (AA: 371-574) expressed in E. Coli. | |
| Primary | |
| 100 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title