missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin S Rabbit anti-Mouse, Clone: 54, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90713-50UL
This item is not returnable.
View return policy
Description
Cathepsin S Monoclonal specifically detects Cathepsin S in Mouse samples. It is validated for ELISA.
Specifications
| Cathepsin S | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1520 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 54 | |
| ELISA 1:5000-1:10000 | |
| cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
| Recombinant Mouse Cathepsin S protein | |
| 50 μg | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction