missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin S Rabbit anti-Mouse, Clone: 54, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 569.00
Specifications
| Antigen | Cathepsin S |
|---|---|
| Clone | 54 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203846
|
Novus Biologicals
NBP2-90713-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18078043
|
Novus Biologicals
NBP2-90713-100UL |
100 μg |
€ 569.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin S Monoclonal specifically detects Cathepsin S in Mouse samples. It is validated for ELISA.Specifications
| Cathepsin S | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 1520 | |
| IgG | |
| Protein A purified |
| 54 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
| Recombinant Mouse Cathepsin S protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title