missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIAS1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94558-0.1ml
This item is not returnable.
View return policy
Description
PIAS1 Polyclonal antibody specifically detects PIAS1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PIAS1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| AR interacting protein, DDXBP1MGC141878, DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1, DEAD/H box-binding protein 1, E3 SUMO-protein ligase PIAS1, GBPRNA helicase II-binding protein, GU/RH-II, Gu-binding protein, MGC141879, Protein inhibitor of activated STAT protein 1, protein inhibitor of activated STAT, 1, protein inhibitor of activated STAT-1, zinc finger, MIZ-type containing 3, ZMIZ3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human PIAS1 (NP_057250.1). MADSAELKQMVMSLRVSELQVLLGYAGRNKHGRKHELLTKALHLLKAGCSPAVQMKIKELYRRRFPQKIMTPADLSIPNVHSSPMPATLSPSTIPQLTYDGHPASSPLLPVSLLGPKHELELPHLTSALHPVHPDIKLQK | |
| 0.1 mL | |
| Signal Transduction | |
| 8554 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction