missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caveolin 2 Recombinant Rabbit Monoclonal Antibody (23GB5020), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552770
This item is not returnable.
View return policy
Description
Caveolae are specialized domains of the plasma membrane that are implicated in the sequestration of a variety of lipid and protein molecules. It has been suggested that these important cellular organelles have a pivotal role in such diverse biochemical processes as lipid metabolism, growth regulation, signal transduction, and apoptosis. Caveolin interacts with and regulates heterotrimeric G-proteins. Currently, there are three members of the caveolin multigene family which are known to encode 21-24 kDa integral membrane proteins that comprise the major structural component of the caveolar membrane in vivo. Caveolin-2 protein is abundantly expressed in fibroblasts and differentiated adipocytes, smooth and skeletal muscle, and endothelial cells. The expression of caveolin-1 is similar to that of caveolin-2 while caveolin-3 expression appears to be limited to muscle tissue types.
Specifications
| Caveolin 2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CAV2 | |
| AI447843; CAV; CAV2; caveolae; caveolae protein, 20-kD; caveolin 2; caveolin 2 isoform a and b; caveolin-2; MGC12294; Phospho-CAV2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12390, 858 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5020 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51636, Q9WVC3 | |
| CAV2 | |
| A synthesized peptide derived from human Caveolin-2 (1-50AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering