missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caveolin 2 Recombinant Rabbit Monoclonal Antibody (23GB5020), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 195.00 - € 468.00
Specifications
| Antigen | Caveolin 2 |
|---|---|
| Clone | 23GB5020 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087842
![]() |
Invitrogen™
MA552770 |
100 μL |
€ 468.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087669
|
Invitrogen™
MA552771 |
20 μL |
€ 195.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caveolin 2 Recombinant Monoclonal antibody specifically detects Caveolin 2 in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Caveolin 2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P51636, Q9WVC3 | |
| 12390, 858 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5020 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI447843; CAV; CAV2; caveolae; caveolae protein, 20-kD; caveolin 2; caveolin 2 isoform a and b; caveolin-2; MGC12294; Phospho-CAV2 | |
| A synthesized peptide derived from human Caveolin-2 (1-50AA). | |
| Primary | |
| -20°C | |
| CAV2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title