missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBL, p Tyr774 Antibody (SR1098), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21601-100ul
This item is not returnable.
View return policy
Description
CBL Monoclonal antibody specifically detects CBL in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| CBL | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| SR1098 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
| Cas-Br-M (murine) ecotropic retroviral transforming sequence, Casitas B-lineage lymphoma proto-oncogene, CBL2E3 ubiquitin-protein ligase CBL, EC 6.3.2, EC 6.3.2.-, oncogene CBL2, Proto-oncogene c-Cbl, RING finger protein 55, RNF55C-CBL, Signal transduction protein CBL | |
| A synthesized peptide derived from human phospho-CBL [Tyr774] (Uniprot #: P22681) | |
| 100 μg | |
| Cancer, Phospho Specific, Signal Transduction, Zinc Finger | |
| 867 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction