missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX4 Antibody (S07-2F2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19821-100ul
This item is not returnable.
View return policy
Description
CBX4 Monoclonal antibody specifically detects CBX4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CBX4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S07-2F2 | |
| Unconjugated | |
| chromobox homolog 4, chromobox homolog 4 (Drosophila Pc class), Chromobox protein homolog 4, Drosophila), E3 SUMO-protein ligase CBX4, hPC2, NS5ATP1-binding protein 16, Pc class 2 homolog, PC2, Polycomb 2 homolog | |
| A synthetic peptide of human CBX4 (Uniprot # O00257) | |
| 100 μg | |
| Apoptosis | |
| 8535 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction