missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX4 Antibody (S07-2F2), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | CBX4 |
|---|---|
| Clone | S07-2F2 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665993
|
Novus Biologicals
NBP3-19821-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608713
|
Novus Biologicals
NBP3-19821-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBX4 Monoclonal antibody specifically detects CBX4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CBX4 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| chromobox homolog 4, chromobox homolog 4 (Drosophila Pc class), Chromobox protein homolog 4, Drosophila), E3 SUMO-protein ligase CBX4, hPC2, NS5ATP1-binding protein 16, Pc class 2 homolog, PC2, Polycomb 2 homolog | |
| A synthetic peptide of human CBX4 (Uniprot # O00257) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S07-2F2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 8535 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title