missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX7 Antibody (3H9C4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61723
This item is not returnable.
View return policy
Description
CBX7 Monoclonal specifically detects CBX7 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| CBX7 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| chromobox homolog 7, chromobox protein homolog 7 | |
| Mouse | |
| 93 kDa | |
| 0.1 mL | |
| 23492 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 3H9C4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CBX7 | |
| Purified recombinant fragment of human CBX7 (AA: 15-147) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction