missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBX7 Antibody (3H9C4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | CBX7 |
|---|---|
| Clone | 3H9C4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18225131
|
Novus Biologicals
NBP2-61723 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638786
|
Novus Biologicals
NBP2-61723-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CBX7 Monoclonal specifically detects CBX7 in Human samples. It is validated for Western Blot, ELISA.Specifications
| CBX7 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 23492 | |
| Purified recombinant fragment of human CBX7 (AA: 15-147) expressed in E. Coli. | |
| Primary | |
| 93 kDa |
| 3H9C4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| chromobox homolog 7, chromobox protein homolog 7 | |
| CBX7 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title