Learn More
CC Chemokine Receptor D6 Antibody
Goat Polyclonal Antibody
Brand:  Novus Biologicals NB100-703-0.025MG
 View more versions of this product
Product Code. 18230115
  • € 194.00 / 25µg
Description
CC Chemokine Receptor D6 Polyclonal specifically detects CC Chemokine Receptor D6 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CC Chemokine Receptor D6
Polyclonal
Unconjugated
10 mM KHPO4, 140 mM NaCl, BSA @ 1mg/ml with 0.1% Sodium Azide
CCBP2
Synthetic peptide SSSIYDYDYLDDMTILVCRKDEVLSFGR corresponding to aa 18-45 of mouse beta chemokine receptor D6
0.025 mg
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
1238
Mouse
IgG
Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Flow Cytometry
2.0 mg/mL
Flow Cytometry 1-5ul/ml, ELISA 1:50000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250
C-C chemokine receptor D6, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250
Goat
Affinity Purified
RUO
Primary
Peptide sequence is < 50 % identical to other mouse chemokine receptors in this region.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.