missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CC Chemokine Receptor D6 Antibody
Goat Polyclonal Antibody
€ 194.00
Specifications
| Antigen | CC Chemokine Receptor D6 |
|---|---|
| Concentration | 2.0 mg/mL |
| Dilution | Flow Cytometry 1-5ul/ml, ELISA 1:50000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250 |
| Applications | Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Flow Cytometry |
| Classification | Polyclonal |
Description
CC Chemokine Receptor D6 Polyclonal specifically detects CC Chemokine Receptor D6 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CC Chemokine Receptor D6 | |
| Flow Cytometry 1-5ul/ml, ELISA 1:50000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250 | |
| Polyclonal | |
| Goat | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 10 mM KHPO4, 140 mM NaCl, BSA @ 1mg/ml with 0.1% Sodium Azide | |
| 1238 | |
| Synthetic peptide SSSIYDYDYLDDMTILVCRKDEVLSFGR corresponding to aa 18-45 of mouse beta chemokine receptor D6 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 2.0 mg/mL | |
| Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Mouse | |
| C-C chemokine receptor D6, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250 | |
| CCBP2 | |
| IgG | |
| Affinity Purified | |
| Peptide sequence is < 50 % identical to other mouse chemokine receptors in this region. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title