missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRAGE/MAGED1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-98411
This item is not returnable.
View return policy
Description
NRAGE/MAGED1 Polyclonal specifically detects NRAGE/MAGED1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| NRAGE/MAGED1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DLXIN-1, MAGE tumor antigen CCF, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen | |
| Rabbit | |
| 86 kDa | |
| 100 μL | |
| Cancer | |
| 9500 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_008917 | |
| MAGED1 | |
| The immunogen for this antibody is MAGED1 - C-terminal region. Peptide sequence ACFVLEKKFGIQLKEIDKEEHLYILISTPESLAGILGTTKDTPKLGLLLV. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Goat: 86%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction