Learn More
CCR7 (CD197) Rat anti-Human, BB700, Clone: 3D12, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566438
This item is not returnable.
View return policy
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR7 (CD197) | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 25 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
| Flow Cytometry | |
| 3D12 | |
| Rat | |
| RUO | |
| Human | |
| Purified |