missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Rat anti-Human, BB700, Clone: 3D12, BD Horizon™
Rat Monoclonal Antibody
€ 178.69 - € 471.28
Specifications
| Antigen | CCR7 (CD197) |
|---|---|
| Clone | 3D12 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15895049
|
BD Biosciences
566438 |
25 Tests |
€ 180.00 € 178.69 / Each Save € 1.31 0.73% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15885049
|
BD Biosciences
566437 |
100 Tests |
€ 472.00 € 471.28 / Each Save € 0.72 0.15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry.Specifications
| CCR7 (CD197) | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Rat | |
| Human | |
| Primary |
| 3D12 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title