Learn More
CCS Mouse, anti-Human, CoraLite Plus 488, Clone: 3F5E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68341-100UL
Description
The CCS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCS. This antibody recognizes human, mouse, rat antigen. The CCS antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CCS | |
| 3F5E5 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence | |
| 1000 μg/mL | |
| CCS | |
| 100 μL | |
| 9973 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |