missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCS Mouse, anti-Human, CoraLite Plus 488, Clone: 3F5E5, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | CCS |
|---|---|
| Clone | 3F5E5 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18077415
|
Proteintech
CL488-68341-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094715
|
Proteintech
CL488-68341-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CCS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCS. This antibody recognizes human, mouse, rat antigen. The CCS antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CCS | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 9973 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3F5E5 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| CCS | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title