Learn More
CCT5 Mouse, anti-Human, CoraLite Plus 488, Clone: 1B12E10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67400-20UL
Beschreibung
The CCT5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CCT5. This antibody recognizes human, mouse, rat antigen. The CCT5 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Spezifikation
| CCT5 | |
| 1B12E10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| CCT epsilon, CCT5, CCTE, KIAA0098, TCP 1 epsilon | |
| 20 μL | |
| 22948 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.